accessibility menu, dialog, popup

UserName

CAS RN 35244-11-2

OHOO

CAS RN 35244-11-2

IUPAC Name: 2-(2-hydroxyphenyl)-4H-chromen-4-one
Synonyms: 2-(2-hydroxyphenyl)chromen-4-one
Molecular Weight (g/mol): 238.24
Molecular Formula: C15H10O3
InChi Key: ZZLQHXCRRMUGQJ-UHFFFAOYSA-N
SMILES: OC1=CC=CC=C1C1=CC(=O)C2=CC=CC=C2O1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)