CAS RN 352535-83-2
IUPAC Name:
(5-chloro-2-fluorophenyl)boronic acid
Synonyms:
5-chloro-2-fluorophenylboronic acid
Molecular Weight (g/mol):
174.36
Molecular Formula:
C6H5BClFO2
InChi Key:
GGTUVWGMCFXUAS-UHFFFAOYSA-N
SMILES:
OB(O)C1=CC(Cl)=CC=C1F
1
–
1
of
1
results
1
5-Chloro-2-fluorobenzeneboronic acid, 97%
CAS: 352535-83-2 Molecular Formula: C6H5BClFO2 Molecular Weight (g/mol): 174.36 MDL Number: MFCD05664225 InChI Key: GGTUVWGMCFXUAS-UHFFFAOYSA-N Synonym: 5-chloro-2-fluorophenyl boronic acid,5-chloro-2-fluorobenzeneboronic acid,5-chloro-2-fluorophenylboronicacid,5-chloro-2-fluoro-phenyl boronic acid,boronic acid, 5-chloro-2-fluorophenyl,pubchem11819,acmc-1ae0a,boronic acid,b-5-chloro-2-fluorophenyl,boronic acid, b-5-chloro-2-fluorophenyl PubChem CID: 3718925 IUPAC Name: (5-chloro-2-fluorophenyl)boronic acid SMILES: OB(O)C1=CC(Cl)=CC=C1F