accessibility menu, dialog, popup

UserName

FFFFFFFFFFFFFF

CAS RN 355-42-0

IUPAC Name: tetradecafluorohexane
Synonyms: perfluorohexane perfluoro-N-hexane n-tetradecafluorohexane perflexane n-perfluorohexane
Molecular Weight (g/mol): 338.04
Molecular Formula: C6F14
InChi Key: ZJIJAJXFLBMLCK-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (5)
  • (3)