missing translation for 'onlineSavingsMsg'
Learn More

Perfluorohexanes (Mixture of Isomers), TRC

Product Code. 30419499
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30419499 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30419499 Supplier Toronto Research Chemicals Supplier No. TRCP9992231G
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perfluorohexanes (Mixture of Isomers)
CAS 355-42-0
Molecular Formula C6F14
Quantity 1 g
Synonym Tetradecafluorohexane, Hexane','1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluoro-, Hexane','tetradecafluoro- (6CI,8CI,9CI), 1,1,1,2,2,3,3,4,4,5,5,6,6,6-Tetradecafluorohexane, AF 0150, FC 72, Fluorinert FC 72, Fluorinert FX 3250D, Fluorinert PF 5060, Flutec PP 1, PF 5060, PFC 51-14, PP 1, Perflexane, Perfluoro-n-hexane, Perfluorohexane, Tetradecafluorohexane, n-Tetradecafluorohexane
InChI Formula InChI=1S/C6F14/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)20
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,6-tetradecafluorohexane
Molecular Weight (g/mol) 338.04
Formula Weight 337.9776
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.