CAS RN 3557-99-1
CAS RN 3557-99-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-(4-Nitrophenyl) 1,4-Benzenedicarboxylate, TRC
CAS: 3557-99-1 Molecular Formula: C14H9NO6 Molecular Weight (g/mol): 287.22 Synonym: p-Nitrophenyl Terephthalic Acid (7CI),mono(4-Nitrophenyl) 1,4-Benzenedicarboxylic Acid (9CI),4-((4-Nitrophenoxy)carbonyl)benzoic Acid IUPAC Name: 4-(4-nitrophenoxy)carbonylbenzoic acid SMILES: OC(=O)c1ccc(cc1)C(=O)Oc2ccc(cc2)[N+](=O)[O-]