missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Nitrophenyl) 1,4-Benzenedicarboxylate, TRC

Product Code. 30425288
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30425288 5 mg 5mg
30414605 50 mg -
30419889 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425288 Supplier Toronto Research Chemicals Supplier No. TRCN8282305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3557-99-1
Molecular Formula C14H9NO6
Molecular Weight (g/mol) 287.22
Synonym p-Nitrophenyl Terephthalic Acid (7CI), mono(4-Nitrophenyl) 1,4-Benzenedicarboxylic Acid (9CI), 4-((4-Nitrophenoxy)carbonyl)benzoic Acid
IUPAC Name 4-(4-nitrophenoxy)carbonylbenzoic acid
SMILES OC(=O)c1ccc(cc1)C(=O)Oc2ccc(cc2)[N+](=O)[O-]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H9NO6/c16-13(17)9-1-3-10(4-2-9)14(18)21-12-7-5-11(6-8-12)15(19)20/h1-8H,(H,16,17)
Formula Weight 287.043
Chemical Name or Material 1-(4-Nitrophenyl) 1,4-Benzenedicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.