accessibility menu, dialog, popup

UserName

BrNN

CAS RN 35590-37-5

IUPAC Name: 5-bromopyridine-3-carbonitrile
Synonyms: 5-bromopyridine-3-carbonitrile
Molecular Weight (g/mol): 183.01
Molecular Formula: C6H3BrN2
InChi Key: FTFFHWWIPOQCBC-UHFFFAOYSA-N
SMILES: BrC1=CN=CC(=C1)C#N
Quantity 
  • (3)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)