missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-cyanopyridine, 97%, Thermo Scientific™

Product Code. 10663205 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
10663205 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 10663205

Brand: Thermo Scientific Acros 399200010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 35590-37-5
Melting Point 104°C to 105°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Molecular Formula C6H3BrN2
MDL Number MFCD00174363
Quantity 1 g
Synonym 5-bromonicotinonitrile, 3-bromo-5-cyanopyridine, 5-bromo-3-cyanopyridine, 3-cyano-5-bromopyridine, 3-pyridinecarbonitrile, 5-bromo, 3-bromo-5-cyano-pyridine, 5-bromo-3-pyridinecarbonitrile, pubchem7702, acmc-209ihz, ksc492g7p
InChI Key FTFFHWWIPOQCBC-UHFFFAOYSA-N
SMILES BrC1=CN=CC(=C1)C#N
IUPAC Name 5-bromopyridine-3-carbonitrile
Molecular Weight (g/mol) 183.01
PubChem CID 736793
Formula Weight 183.01
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-cyanopyridine
Show More Show Less
compliance-icons
Product Identifier
  • 5-Bromo-3-cyanopyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.