accessibility menu, dialog, popup

UserName

CH3OONH2OH

CAS RN 35748-34-6

IUPAC Name: methyl 3-amino-2-hydroxybenzoate
Synonyms: methyl 3-amino-2-hydroxybenzoate
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: OMWQHVRUXLRZRC-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(O)C(N)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results