accessibility menu, dialog, popup

UserName

HN(S)NHOOONHH2NOHCH3CH3H3CHCl

CAS RN 35897-34-8

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-5-[(diaminomethylidene)amino]pentanoic acid
Synonyms: 2-[(tert-butoxycarbonyl)amino]-5-[(diaminomethylidene)amino]pentanoic acid
Molecular Weight (g/mol): 274.32
Molecular Formula: C11H22N4O4
InChi Key: HSQIYOPBCOPMSS-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCCN=C(N)N)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)