missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-Boc-L-arginine hydrochloride, 98%

Product Code. 15490389
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490389 25 g 25g
15480389 5 g 5g
15400399 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490389 Supplier Thermo Scientific Alfa Aesar Supplier No. H63611.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35897-34-8 | C11H23ClN4O4 | 310.78 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35897-34-8
Molecular Formula C11H23ClN4O4
Molecular Weight (g/mol) 310.78
MDL Number MFCD00065552
InChI Key HDELGKMVZYHPPB-RZUXQYDTNA-N
Synonym boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, boc-arg-oh.hcl.h2o, boc-d-arg-oh hcl h2o, boc-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932, boc-l-arg-ohhclh2o
PubChem CID 2729004
SMILES Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O

Specifications

Color White
Beilstein 3658939
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate;hydrochloride
Quantity 25 g
Formula Weight 310.78
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Nalpha-Boc-L-arginine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.