CAS RN 361154-30-5
CAS RN 361154-30-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ Ac4ManNAz
Azide-containing monosaccharide building block; used as glycoprotein, glycolipid and glycoRNA probe