accessibility menu, dialog, popup

UserName

CAS RN 3612-20-2

IUPAC Name: 1-benzylpiperidin-4-one
Synonyms: 1-benzylpiperidin-4-one
Molecular Weight (g/mol): 189.26
Molecular Formula: C12H15NO
InChi Key: SJZKULRDWHPHGG-UHFFFAOYSA-N
SMILES: O=C1CCN(CC2=CC=CC=C2)CC1
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)