Learn More
1-Benzyl-4-piperidone, 98+%
Description
1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Benzyl-4-piperidone is a pharmaceutical intermediate, it is used in penfluridol synthesis of bulk drugs.
Solubility
Soluble in water (12 g/100ml at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Spécification
Spécification
| Density | 1.059 |
| Boiling Point | 248°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.5399 |
| Quantity | 500 g |
| Beilstein | 128556 |
| Solubility Information | Soluble in water (12g/100ml at 20°C). |
| Formula Weight | 189.26 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 1-Benzyl-4-piperidone |
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.