accessibility menu, dialog, popup

UserName

CH3OBrOCH3

CAS RN 36255-44-4

IUPAC Name: 3-bromo-1,1-dimethoxypropane
Synonyms: 3-bromo-1,1-dimethoxypropane
Molecular Weight (g/mol): 183.05
Molecular Formula: C5H11BrO2
InChi Key: ODZZAIFAQLODKN-UHFFFAOYSA-N
SMILES: COC(CCBr)OC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results