missing translation for 'onlineSavingsMsg'
Learn More

3-bromopropionaldehyde dimethylacetal, 95%, Thermo Scientific Chemicals

Product Code. 10044342 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10044342 5 g Glass bottle 5g
10240543 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10044342 Supplier Thermo Scientific Acros Supplier No. 376260050
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36255-44-4
Molecular Formula C5H11BrO2
Molecular Weight (g/mol) 183.045
MDL Number MFCD00013245
InChI Key ODZZAIFAQLODKN-UHFFFAOYSA-N
Synonym 3-bromopropionaldehyde dimethyl acetal, 1-bromo-3,3-dimethoxypropane, propane, 3-bromo-1,1-dimethoxy, 3-bromo-1,1-dimethoxy-propane, acmc-209ilq, 3,3-dimethoxypropyl bromide, 3-bromopropanal dimethylacetal, ksc495q3l, 3-bromo-1,1dimethoxy-propane, 1,1-dimethoxy-3-bromopropane
PubChem CID 118932
IUPAC Name 3-bromo-1,1-dimethoxypropane
SMILES COC(CCBr)OC

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.3410g/mL
Boiling Point 58.0°C to 60.0°C (14.0 mmHg)
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula Br(CH2)2CH(OCH3)2
Refractive Index 1.4460 to 1.4500 (20°C, 589nm)
Quantity 5 g
Formula Weight 183.04
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Bromopropionaldehyde dimethylacetal
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromopropionaldehyde dimethylacetal
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.