accessibility menu, dialog, popup

UserName

H3CSOONHN

CAS RN 36268-67-4

IUPAC Name: N-(4-cyanophenyl)methanesulfonamide
Synonyms: N-(4-cyanophenyl)methanesulfonamide
Molecular Weight (g/mol): 196.22
Molecular Formula: C8H8N2O2S
InChi Key: BRDHOCVMWSXEHI-UHFFFAOYSA-N
SMILES: CS(=O)(=O)NC1=CC=C(C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results