accessibility menu, dialog, popup

UserName

ONOBrF

CAS RN 364-73-8

IUPAC Name: 4-bromo-1-fluoro-2-nitrobenzene
Synonyms: 4-bromo-1-fluoro-2-nitrobenzene
Molecular Weight (g/mol): 220
Molecular Formula: C6H3BrFNO2
InChi Key: UQEANKGXXSENNF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Br)=CC=C1F
Boiling Point 
  • (5)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results