accessibility menu, dialog, popup

UserName

CAS RN 3641-14-3

CH3OONHNH2NN

CAS RN 3641-14-3

IUPAC Name: methyl 3-amino-1H-1,2,4-triazole-5-carboxylate
Synonyms: methyl 5-amino-2H-1,2,4-triazole-3-carboxylate
Molecular Weight (g/mol): 142.12
Molecular Formula: C4H6N4O2
InChi Key: OSZNPKRMPBUQLB-UHFFFAOYSA-N
SMILES: COC(=O)C1=NC(N)=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results