missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-1H-[1,2,4]-triazole-3-carboxylic acid methyl ester, 96%, Thermo Scientific™

Product Code. 10473520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10473520 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10473520 Supplier Thermo Scientific Acros Supplier No. 437690010
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 3641-14-3
CAS Min % 95.0
CAS Max % 100.0
Melting Point 212°C to 213°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Molecular Formula C4H6N4O2
Quantity 1 g
Synonym methyl 5-amino-1h-1,2,4-triazole-3-carboxylate, methyl 5-amino-4h-1,2,4-triazole-3-carboxylate, methyl 5-amino-4h-1,2,4 triazole-3-carboxylate, 1h-1,2,4-triazole-3-carboxylic acid, 5-amino-, methyl ester, 5-amino-1h-1,2,4-triazole-3-carboxylic acid methyl ester, methyl 5-amino-1,2,4-triazole-3-carboxylate, methyl-5-amino-1h-1,2,4-triazole-3-carboxylate, 5-amino-1h-1,2,4 triazole-3-carboxylic acid methyl ester, methyl 5-amino-2h-1,2,4-triazole-3-carboxylate
InChI Key OSZNPKRMPBUQLB-UHFFFAOYSA-N
SMILES COC(=O)C1=NC(=NN1)N
IUPAC Name methyl 3-amino-1H-1,2,4-triazole-5-carboxylate
Molecular Weight (g/mol) 142.12
PubChem CID 77201
Formula Weight 142.12
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 5-Amino-1H-[1,2,4]-triazole-3-carboxylic acid methyl ester
Show More Show Less
compliance-icons
Product Identifier
  • 5-Amino-1H-[1,2,4]-triazole-3-carboxylic acid methyl ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.