accessibility menu, dialog, popup

UserName

CAS RN 365-24-2

HOFF

CAS RN 365-24-2

IUPAC Name: bis(4-fluorophenyl)methanol
Synonyms: bis(4-fluorophenyl)methanol
Molecular Weight (g/mol): 220.22
Molecular Formula: C13H10F2O
InChi Key: WCTZPQWLFWZYJE-UHFFFAOYSA-N
SMILES: OC(C1=CC=C(F)C=C1)C1=CC=C(F)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results