accessibility menu, dialog, popup

UserName

CH3OONH3C

CAS RN 3652-61-7

IUPAC Name: ethyl 2-methyl-1,5-diphenyl-1H-pyrrole-3-carboxylate
Synonyms: ethyl 2-methyl-1,5-diphenylpyrrole-3-carboxylate
Molecular Weight (g/mol): 305.38
Molecular Formula: C20H19NO2
InChi Key: YHEGUULWFOVVRG-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N(C(=C1)C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results