accessibility menu, dialog, popup

UserName

BrOO

CAS RN 36603-49-3

IUPAC Name: 2-(4-bromophenoxy)oxane
Synonyms: 2-(4-bromophenoxy)oxane
Molecular Weight (g/mol): 257.13
Molecular Formula: C11H13BrO2
InChi Key: MXDQGXMBJCGRCB-UHFFFAOYNA-N
SMILES: BrC1=CC=C(OC2CCCCO2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results