missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenoxy)tetrahydropyran, 98%

Product Code. 11495660
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495660 50 g 50g
11485660 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11495660 Supplier Thermo Scientific Alfa Aesar Supplier No. H25914.18
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 36603-49-3 | C11H13BrO2 | 257.127 g/mol

2-(4-Bromophenoxy)tetrahydropyran can be used to produce 4-bromo-phenol at temperature of 20°C. It is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Bromophenoxy)tetrahydropyran can be used to produce 4-bromo-phenol at temperature of 20°C. It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36603-49-3
Molecular Formula C11H13BrO2
Molecular Weight (g/mol) 257.127
MDL Number MFCD00091551
InChI Key MXDQGXMBJCGRCB-UHFFFAOYSA-N
Synonym 2-4-bromophenoxy tetrahydro-2h-pyran, 2-4-bromophenoxy tetrahydropyran, 2-4-bromophenoxy oxane, 2h-pyran, 2-4-bromophenoxy tetrahydro, 2-4'-bromophenoxy-tetrahydropyran, 4-bromophenyl thp ether, acmc-209inn, bromophenoxy tetrahydro-2h-pyran, 2-4-bromophenoxy-tetrahydropyran
PubChem CID 4646436
IUPAC Name 2-(4-bromophenoxy)oxane
SMILES C1CCOC(C1)OC2=CC=C(C=C2)Br

Specifications

Melting Point 55°C to 58°C
Quantity 50 g
Solubility Information Slightly soluble in water.
Formula Weight 257.13
Percent Purity 98%
Chemical Name or Material 2-(4-Bromophenoxy)tetrahydropyran

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.