CAS RN 36669-06-4
CAS RN 36669-06-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dimethyl 2-Hydroxyisophthalate, TRC
CAS: 36669-06-4 Molecular Formula: C10H10O5 Molecular Weight (g/mol): 210.18 Synonym: 2-Hydroxy-1,3-benzenedicarboxylic Acid Dimethyl Ester,2,6-(Dicarbomethoxy)phenol,2-Hydroxyisophthalic Acid Dimethyl Ester,Dimethyl 2-Hydroxy-1,3-benzenedicarboxylate SMILES: COC(=O)c1cccc(C(=O)OC)c1O