missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl 2-Hydroxyisophthalate, TRC

Product Code. 30388453
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388453 1 g
30377181 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388453 Supplier Toronto Research Chemicals Supplier No. TRCD4732601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36669-06-4
Molecular Formula C10H10O5
Molecular Weight (g/mol) 210.18
Synonym 2-Hydroxy-1,3-benzenedicarboxylic Acid Dimethyl Ester, 2,6-(Dicarbomethoxy)phenol, 2-Hydroxyisophthalic Acid Dimethyl Ester, Dimethyl 2-Hydroxy-1,3-benzenedicarboxylate
SMILES COC(=O)c1cccc(C(=O)OC)c1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H10O5/c1-14-9(12)6-4-3-5-7(8(6)11)10(13)15-2/h3-5,11H,1-2H3
Formula Weight 210.0528
Chemical Name or Material Dimethyl 2-Hydroxyisophthalate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.