accessibility menu, dialog, popup

UserName

CAS RN 368-66-1

IUPAC Name: tris(trifluoromethyl)-1,3,5-triazine
Synonyms: tris(trifluoromethyl)-1,3,5-triazine
Molecular Weight (g/mol): 285.07
Molecular Formula: C6F9N3
InChi Key: LSGBKABSSSIRJF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=NC(=NC(=N1)C(F)(F)F)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)