missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-tris(Trifluoromethyl)-1,3,5-triazine, TRC

Product Code. 30136655
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136655 1 g 1g
17980855 100 mg 100mg
17903985 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136655 Supplier Toronto Research Chemicals Supplier No. TRCT7952281G
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-66-1
Molecular Formula C6N3F9
Molecular Weight (g/mol) 285.06
Synonym 2,4,6-Tris(trifluoromethyl)-s-triazine
IUPAC Name 2,4,6-tris(trifluoromethyl)-1,3,5-triazine
SMILES FC(F)(F)c1nc(nc(n1)C(F)(F)F)C(F)(F)F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6F9N3/c7-4(8,9)1-16-2(5(10,11)12)18-3(17-1)6(13,14)15
Formula Weight 284.9949
Chemical Name or Material 2,4,6-tris(Trifluoromethyl)-1,3,5-triazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.