accessibility menu, dialog, popup

UserName

NH2FF

CAS RN 368-99-0

IUPAC Name: 3-(difluoromethyl)aniline
Synonyms: 3-(difluoromethyl)aniline
Molecular Weight (g/mol): 143.14
Molecular Formula: C7H7F2N
InChi Key: IDFPXKDJNDYDKA-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results