missing translation for 'onlineSavingsMsg'
Learn More

3-(Difluoromethyl)aniline, 97%

Product Code. 15487117
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15487117 250 mg 250mg
15477117 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15487117 Supplier Thermo Scientific Alfa Aesar Supplier No. H34188.MD
 Restricted Item
Estimated Shipment: 12-10-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.

CAS: 368-99-0 | C7H7F2N | 143.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 368-99-0
Molecular Formula C7H7F2N
Molecular Weight (g/mol) 143.14
MDL Number MFCD18384830
InChI Key IDFPXKDJNDYDKA-UHFFFAOYSA-N
Synonym 3-difluoromethyl aniline, 3-difluoromethyl benzenamine, 3-difluoromethyl-phenylamine, 3-difluoromethyl benzenamine hcl
PubChem CID 23445119
IUPAC Name 3-(difluoromethyl)aniline
SMILES NC1=CC=CC(=C1)C(F)F

Tekniske data

Boiling Point 82°C (4 mmHg)
Quantity 250 mg
UN Number UN2810
Formula Weight 143.14
Percent Purity 97%
Chemical Name or Material 3-(Difluoromethyl)aniline

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.