accessibility menu, dialog, popup

UserName

NH2SNHCl

CAS RN 3696-23-9

IUPAC Name: (4-chlorophenyl)thiourea
Synonyms: 1-(O-chlorophenyl)-2-thiourea
Molecular Weight (g/mol): 186.66
Molecular Formula: C7H7ClN2S
InChi Key: XVEFWRUIYOXUGG-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results