accessibility menu, dialog, popup

UserName

CH3SOOBOHHO

CAS RN 373384-18-0

IUPAC Name: (3-methanesulfonylphenyl)boronic acid
Synonyms: 3-methanesulfonylphenylboronic acid
Molecular Weight (g/mol): 200.02
Molecular Formula: C7H9BO4S
InChi Key: HZFFUMBZBGETES-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=CC(=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results