accessibility menu, dialog, popup

UserName

HHCHCClZr

CAS RN 37342-97-5

IUPAC Name: λ²-zirconium(2+) bis(cyclopenta-2,4-dien-1-ide) chloride hydride
Synonyms: λ²-zirconium(2+) bis(cyclopentadienide) chloride hydride
Molecular Weight (g/mol): 257.87
Molecular Formula: C10H11ClZr
InChi Key: VPQMECJLNPUNGN-UHFFFAOYSA-M
SMILES: [H-].[Cl-].[Zr++].[CH-]1C=CC=C1.[CH-]1C=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)