missing translation for 'onlineSavingsMsg'
Learn More

Bis(cyclopentadienyl)zirconium chloride hydride, 95%

Product Code. 10762315 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10762315 25 g Glass bottle 25g
10391234 1 g Glass bottle 1g
10236790 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10762315 Supplier Thermo Scientific Acros Supplier No. 207890250
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 37342-97-5 | C10H11ClZr | 257.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37342-97-5
Molecular Formula C10H11ClZr
Molecular Weight (g/mol) 257.86
MDL Number MFCD02089401
InChI Key VPQMECJLNPUNGN-UHFFFAOYSA-M
Synonym zirconocene chloride hydride
PubChem CID 131718033
IUPAC Name cyclopenta-1,3-diene;hydride;zirconium(2+);chloride
SMILES [H-].[CH-]1C=CC=C1.[CH-]1C=CC=C1.[Cl-].[Zr+2]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point >300.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 8533
Solubility Information Solubility in water: reacts with water.
Formula Weight 257.86
Percent Purity 95%
Physical Form Powder
Chemical Name or Material Bis(cyclopentadienyl)zirconium chloride hydride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Bis(cyclopentadienyl)zirconium chloride hydride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 1 B
  • Substance which, in contact with water, emits flammable gases Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H261-In contact with water releases flammable gases.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.