CAS RN 37348-16-6
CAS RN 37348-16-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hydrogen bromide, 33% w/w (45% w/v) soln. in acetic acid
CAS: 37348-16-6 Molecular Formula: BrH Molecular Weight (g/mol): 80.91 MDL Number: MFCD00011323 InChI Key: CPELXLSAUQHCOX-UHFFFAOYSA-N Synonym: hydrogen bromide,hydrobromic acid,bromwasserstoff,broomwaterstof,bromowodor,acido bromidrico,acide bromhydrique,anhydrous hydrobromic acid,bromowodor polish,hydrobromide PubChem CID: 260 ChEBI: CHEBI:47266 IUPAC Name: bromane SMILES: Br