accessibility menu, dialog, popup

UserName

CAS RN 374105-86-9

IUPAC Name: (2E)-3-[2-(dihydroxyboranyl)phenyl]prop-2-enoic acid
Synonyms: (2E)-3-[2-(dihydroxyboranyl)phenyl]prop-2-enoic acid
Molecular Weight (g/mol): 191.98
Molecular Formula: C9H9BO4
InChi Key: OPFDGNUJQALQFL-AATRIKPKSA-N
SMILES: OB(O)C1=CC=CC=C1\C=C\C(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results