accessibility menu, dialog, popup

UserName

CAS RN 374537-97-0

IUPAC Name: 5-bromo-3,4-dimethylpyridin-2-amine
Synonyms: 5-bromo-3,4-dimethylpyridin-2-amine
Molecular Weight (g/mol): 201.07
Molecular Formula: C7H9BrN2
InChi Key: YAVKJNIMFGZBSY-UHFFFAOYSA-N
SMILES: CC1=C(C)C(N)=NC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results