missing translation for 'onlineSavingsMsg'
Learn More

2-Amino-5-bromo-3,4-dimethylpyridine, 97%

Product Code. 11398616
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11398616 5 g 5g
11388616 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398616 Supplier Thermo Scientific Alfa Aesar Supplier No. L17672.06
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 374537-97-0 | C7H9BrN2 | 201.07 g/mol

It is an active pharmaceutical ingredient.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an active pharmaceutical ingredient.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 374537-97-0
Molecular Formula C7H9BrN2
Molecular Weight (g/mol) 201.07
MDL Number MFCD02093947
InChI Key YAVKJNIMFGZBSY-UHFFFAOYSA-N
Synonym 2-amino-5-bromo-3,4-dimethylpyridine, 5-bromo-3,4-dimethyl-2-pyridinamine, acmc-20agep, 5-bromo-3,4-dimethyl-2-pyridylamine, 2-pyridinamine,5-bromo-3,4-dimethyl, 5-bromo-3,4-dimethyl-pyridin-2-ylamine
PubChem CID 7010307
IUPAC Name 5-bromo-3,4-dimethylpyridin-2-amine
SMILES CC1=C(C)C(N)=NC=C1Br

Specifications

Melting Point 133°C to 135°C
Quantity 5 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 201.07
Percent Purity 97%
Chemical Name or Material 2-Amino-5-bromo-3,4-dimethylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.