accessibility menu, dialog, popup

UserName

CH3OOBOHHO

CAS RN 374538-03-1

IUPAC Name: [2-(methoxycarbonyl)phenyl]boronic acid
Synonyms: 2-(methoxycarbonyl)phenylboronic acid
Molecular Weight (g/mol): 179.97
Molecular Formula: C8H9BO4
InChi Key: ODAXNYMENLFYMY-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC=C1B(O)O
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results