missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxycarbonylphenylboronic acid, 97%

Product Code. 10732962 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10732962 5 g Glass bottle 5g
10077949 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Questo articolo non è restituibile. Consulta la politica di reso
Codice prodotto. 10732962 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 433910050
 Restricted Item
Estimated Shipment: 17-11-2026
Aggiungi al carrello
Aggiungi al carrello
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Questo articolo non è restituibile. Consulta la politica di reso

CAS: 374538-03-1 | C8H9BO4 | 179.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 374538-03-1
Molecular Formula C8H9BO4
Molecular Weight (g/mol) 179.97
MDL Number MFCD02179452
InChI Key ODAXNYMENLFYMY-UHFFFAOYSA-N
Synonym 2-methoxycarbonyl phenylboronic acid, 2-methoxycarbonyl benzeneboronic acid, 2-methoxycarbonyl phenyl boronic acid, 2-methoxycarbonylphenyl boronic acid, methyl 2-boronobenzoate, methyl 2-dihydroxyboranyl benzoate, o-methoxycarbonyl phenylboronic acid, 2-carbomethoxybenzeneboronic acid, 2-methoxycarbonylphenylboronicacid
PubChem CID 2773496
SMILES COC(=O)C1=CC=CC=C1B(O)O

Specifica

CAS Min % 96.0
CAS Max % 100.0
Melting Point 101.1°C to 101.4°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
IUPAC Name (2-methoxycarbonylphenyl)boronic acid
Formula Weight 179.97
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methoxycarbonylphenylboronic acid
Vedi altri risultati Mostra meno risultati
compliance-icons
Identificativo prodotto
  • 2-Methoxycarbonylphenylboronic acid, 97%
Avvertenza
  • Warning
Categoria di pericolo
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicazioni di pericolo
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Consigli di prudenza
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.