accessibility menu, dialog, popup

UserName

CAS RN 374564-34-8

BBrFFFK

CAS RN 374564-34-8

IUPAC Name: potassium (3-bromophenyl)trifluoroboranuide
Synonyms: potassium (3-bromophenyl)trifluoroboranuide
Molecular Weight (g/mol): 262.91
Molecular Formula: C6H4BBrF3K
InChi Key: NIDYRVJZTADCGO-UHFFFAOYSA-N
SMILES: [K+].F[B-](F)(F)C1=CC(Br)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results