accessibility menu, dialog, popup

UserName

FFFSOONSOOFFF

CAS RN 37595-74-7

IUPAC Name: 1,1,1-trifluoro-N-phenyl-N-trifluoromethanesulfonylmethanesulfonamide
Synonyms: 1,1,1-trifluoro-N-phenyl-N-trifluoromethanesulfonylmethanesulfonamide
Molecular Weight (g/mol): 357.24
Molecular Formula: C8H5F6NO4S2
InChi Key: DIOHEXPTUTVCNX-UHFFFAOYSA-N
SMILES: FC(F)(F)S(=O)(=O)N(C1=CC=CC=C1)S(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)