accessibility menu, dialog, popup

UserName

CAS RN 376-72-7

HOOFFFFFFFF

CAS RN 376-72-7

IUPAC Name: 2,2,3,3,4,4,5,5-octafluoropentanoic acid
Synonyms: 2,2,3,3,4,4,5,5-octafluoropentanoic acid
Molecular Weight (g/mol): 246.06
Molecular Formula: C5H2F8O2
InChi Key: VGFKXVSMDOKOJZ-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)F
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)