missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-H-Octafluoropentanoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 5-H-Octafluoropentanoic Acid |
| CAS | 376-72-7 |
| Molecular Formula | C5 H2 F8 O2 |
| Quantity | 5 g |
| Synonym | 2,2,3,3,4,4,5,5-Octafluoropentanoic Acid, 2,2,3,3,4,4,5,5,-Octafluoropentanoic Acid, 5-H-Octafluoropentanoic Acid, ω-Hydrooctafluorovaleric Acid, ω-Hydroperfluorovaleric Acid, ω-Monohydrooctafluoropentanoic Acid |
| InChI Formula | InChI=1S/C5H2F8O2/c6-1(7)3(8,9)5(12,13)4(10,11)2(14)15/h1H,(H,14,15) |
| SMILES | OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)F |
| IUPAC Name | 2,2,3,3,4,4,5,5-octafluoropentanoic acid |
| Molecular Weight (g/mol) | 246.055 |
| Formula Weight | 245.993 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?