accessibility menu, dialog, popup

UserName

CAS RN 37778-99-7

IUPAC Name: 2-phenylpropan-1-ol
Synonyms: 2-phenylpropanol-1 hydratropic alcohol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: RNDNSYIPLPAXAZ-UHFFFAOYNA-N
SMILES: CC(CO)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results