missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Phenyl-1-propanol, 99+%, Thermo Scientific™

Product Code. 10627811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
Packaging:
Glass bottle
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10627811 1 mL Glass bottle 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10627811 Supplier Thermo Scientific Acros Supplier No. 264710010
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 37778-99-7
CAS Min % 99.0
CAS Max % 100.0
Color Colorless
Boiling Point 214°C
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C9H12O
Linear Formula C6H5CH(CH3)CH2OH
Refractive Index 1.5252 to 1.5272
MDL Number MFCD00145249
Quantity 1 mL
Specific Rotation Condition −18° (20°C c=neat)
Specific Rotation -18
Synonym s-2-phenylpropan-1-ol, s---2-phenyl-1-propanol, --2-phenylpropanol, s-2-phenyl-1-propanol, 2s-2-phenylpropan-1-ol, unii-w7q89429zo, 2 s-phenyl-1-propanol, --2-phenyl-1-propanol, --beta-methylbenzeneethanol
InChI Key RNDNSYIPLPAXAZ-MRVPVSSYSA-N
SMILES CC(CO)C1=CC=CC=C1
IUPAC Name (2S)-2-phenylpropan-1-ol
Molecular Weight (g/mol) 136.19
PubChem CID 447661
Formula Weight 136.19
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Phenyl-1-propanol, 99+%
Show More Show Less
compliance-icons
Product Identifier
  • (S)-(-)-2-Phenyl-1-propanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.