accessibility menu, dialog, popup

UserName

OHNN

CAS RN 3785-90-8

IUPAC Name: 2-[(4-hydroxyphenyl)methylidene]propanedinitrile
Synonyms: tyrphostin 8
Molecular Weight (g/mol): 170.17
Molecular Formula: C10H6N2O
InChi Key: FNCOVSWSZZVFBQ-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C(C#N)C#N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results