accessibility menu, dialog, popup

UserName

OOH

CAS RN 37868-26-1

IUPAC Name: 2-(2,3-dihydro-1H-inden-2-yl)acetic acid
Synonyms: 2,3-dihydro-1H-inden-2-ylacetic acid
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: TULDPXYHBFBRGW-UHFFFAOYSA-N
SMILES: OC(=O)CC1CC2=CC=CC=C2C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results