CAS RN 37921-38-3
CAS RN 37921-38-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cimifugin, MedChemExpress
MedChemExpress Cimifugin (Cimitin) is a bioactive component of Saposhnikovia divaricata, a Chinese herb. Cimifugin suppresses allergic inflammation by reducing epithelial derived initiative key factors via regulating tight junctions.