accessibility menu, dialog, popup

UserName

CAS RN 38002-45-8

CH3SiCH3CH3Br

CAS RN 38002-45-8

IUPAC Name: (3-bromoprop-1-yn-1-yl)trimethylsilane
Synonyms: (3-bromoprop-1-yn-1-yl)trimethylsilane
Molecular Weight (g/mol): 191.14
Molecular Formula: C6H11BrSi
InChi Key: GAPRPFRDVCCCHR-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CCBr
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)